Learn More
Grainger Power Correction Capacitor 10 kvar
Supplier:  Grainger 16Y171
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y171 is a single-phase capacitor designed to improve power factor in AC power systems. This capacitor has a capacitance of 25 microfarads with a voltage rating of 370 VAC. It can handle currents up to 135 Amps and has a kVAR rating of 7.9. The capacitor uses film and foil technology in a metal enclosed style and is UL listed. It measures 10.25" H x 8.75" W x 8.25" D and weighs approximately 27 lbs. This capacitor helps reduce energy waste and improves voltage regulation in motors, transformers, lighting and other inductive loads. It is well-suited for industrial and commercial applications.
Catalog No. 50-276-5074