Learn More
Grainger Power Correction Capacitor 12.5 kvar
Supplier:  Grainger 16Y172
Encompass_Preferred
The Grainger Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in inductive circuits. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor features a metal enclosure and quick-connect terminals for easy installation. It can help reduce energy costs and improve power quality by correcting power factor issues. This capacitor is UL and CSA listed.
Catalog No. 50-276-5500