Learn More
Grainger Power Correction Capacitor 15 kvar
Supplier:  Grainger 16Y153
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has UL and CSA certifications. The capacitor uses dry polypropylene film dielectric material in a sealed steel enclosure for durability. It can help reduce utility costs and improve voltage conditions when properly applied to inductive loads. This capacitor measures 10.25 x 6 x 12.25 inches and weighs 26 pounds. It is well-suited for use in commercial and industrial facilities.
Catalog No. 50-276-6225