Learn More
Grainger Power Correction Capacitor 17.5 kvar
Supplier:  Grainger 16Y198
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 25 microfarads with a voltage rating of 440VAC. It features an internal discharge resistor for safety and has compact dimensions of 5.9" diameter x 7.5" height. The capacitor casing is made of steel with porcelain insulators. It can provide 22.5kVAR of reactive power compensation and is UL listed. The capacitor helps optimize power distribution systems by reducing line losses and improving voltage regulation. Its high quality construction makes it suitable for industrial and commercial applications.
Catalog No. 50-276-5543