Learn More
Grainger Power Correction Capacitor 17.5 kvar
Supplier:  Grainger 16Y174
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a metal enclosure and is UL and CSA certified. The capacitor helps reduce energy costs and improve voltage conditions by correcting power factor issues. With dimensions of 7"L x 3.5"W x 5.25"H, this 30 microfarad correction capacitor provides an economical solution for power factor correction in commercial and industrial applications.
Catalog No. 50-276-6982