Learn More
Grainger Power Correction Capacitor 2.5 kvar
Supplier:  Grainger 16Y147
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The dimensions are 7" diameter x 17" height and it weighs approximately 35 lbs. This capacitor helps reduce energy consumption and improves voltage regulation when properly applied to inductive loads. Its high quality components ensure reliable performance and long service life.
Catalog No. 50-276-6183