Learn More
Grainger Power Correction Capacitor 25 kvar
Supplier:  Grainger 16Y177
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA approved. The dimensions are 3-1/2" diameter x 10-1/2" height. This capacitor helps reduce energy costs and prevent overheating and premature motor failure by correcting power factor issues. It is well-suited for use with motors, transformers, ballasts, and other inductive loads.
Catalog No. 50-276-5199