Learn More
Grainger Power Correction Capacitor 25 kvar
Supplier:  Grainger 16Y202
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor is enclosed in a steel enclosure with epoxy resin finish for protection. It can operate in temperatures from -25°C to 55°C and has UL and CSA certifications. The dimensions are 7"H x 5"W x 5.5"D. This capacitor helps reduce energy costs and improves power quality in lighting, motor, and other inductive load applications. It is well-suited for use in commercial and industrial facilities.
Catalog No. 50-276-6828