Learn More
Grainger Power Correction Capacitor 27.5 kvar
Supplier:  Grainger 16Y157
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor helps reduce energy waste and lower utility costs by correcting power factor issues. This model has an enclosure rating of NEMA 1 and includes pressure disconnecting devices for safety. With dimensions of 7"H x 6"W x 5"D, this capacitor provides an economical solution for power factor correction in commercial and industrial applications.
Catalog No. 50-276-5061