Learn More
Grainger Power Correction Capacitor 35 kvar
Supplier:  Grainger 16Y180
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y180 is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 180 microfarads with a voltage rating of 480VAC. It features an internal discharge resistor for safety and has an expected life of 100,000 hours at rated voltage and temperature. The capacitor case is made of steel with epoxy resin end bells. Dimensions are 10.2" diameter x 17.7" length. This capacitor weighs 85 lbs and has 3/4"-16 threaded inserts for mounting. It can help reduce energy costs and improve voltage regulation when properly installed in parallel with inductive loads.
Catalog No. 50-276-7816