Learn More
Grainger Power Correction Capacitor 40 kvar
Supplier:  Grainger 16Y181
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y181 is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 30 microfarads with a voltage rating of 480VAC. It features an internal discharge resistor for safety and has an expected life of 100,000 hours at rated voltage and temperature. The capacitor case is made of steel with epoxy resin end bells. Dimensions are 7.4" diameter x 17.3" length and it weighs approximately 44 lbs. This capacitor is UL and CSA listed and ideal for improving power factor in commercial and industrial applications.
Catalog No. 50-276-5195