Learn More
Grainger Power Correction Capacitor 50 kvar
Supplier:  Grainger 16Y183
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 25 microfarads with a voltage rating of 440VAC. It features an internal discharge resistor for safety and has compact dimensions of 5.9"x5.9"x7.9". The capacitor casing is made of steel with epoxy resin insulation between plates. It can provide automatic power factor correction when used with appropriate controls. This capacitor aims to reduce energy waste and improve electrical efficiency.
Catalog No. 50-276-6854