Learn More
Grainger Power Correction Capacitor 50 kvar
Supplier:  Grainger 16Y208
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor helps reduce energy waste and lower utility costs by correcting power factor issues. This model has an enclosed metal case and is UL/CSA recognized. It can be used in commercial and industrial applications to optimize electrical efficiency. The dimensions are 7" x 7" x 12" and it weighs approximately 35 lbs.
Catalog No. 50-276-7796