Learn More
Grainger Power Correction Capacitor 60 kvar
Supplier:  Grainger 16Y209
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480V voltage. It features an internal discharge resistor for safety and has an epoxy resin filled aluminum can type construction. The operating temperature range is -25°C to 55°C. This capacitor measures 18" H x 14" W x 12" D and weighs approximately 88 lbs. It can help reduce energy consumption and improve voltage conditions when properly installed in parallel with inductive loads. This product carries a 12 month warranty from WEG.
Catalog No. 50-276-5542