Learn More
Grainger Power Correction Capacitor 7.5 kvar
Supplier:  Grainger 16Y150
Encompass_Preferred
The Grainger Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in inductive circuits. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor features a metal enclosure and quick-connect terminals for easy installation. It can help reduce utility costs and improve power quality by correcting power factor issues. With dimensions of 4-1/2"H x 3-1/2"W x 3-1/2"D, this capacitor provides an economical solution for power factor correction in commercial and industrial applications.
Catalog No. 50-276-5503