Learn More
Hoefer, Inc Hoefer™ PS600 600 Volt Power Supply
SDP
Supplier:  Hoefer, Inc PS600
Encompass_Preferred
The midrange PS600 is designed for electrophoresis and blotting techniques including large format and high throughput applicationsFeatures:Constant voltage or constant current mode with automatic crossover5 to 600 V, 3 to 750 mA, 150 WFour sets of 4 mm output jacksIdeal choice for most electrophoresis techniquesContinuous runs or 999 minute timerAutomatic recovery after power failureOpen circuit detectionShort circuit protectionModes of Operation:Constant Voltage Mode—the power supply will attempt to maintain the user selected voltage by automatically adjusting the current flowConstant Current Mode—the power supply will attempt to maintain the user selected current flow by automatically adjusting the voltage suppliedAutomatic Crossover—when the constant parameter (voltage or current) cannot be maintained due to power supply operating limits the power supply automatically switches to maintaining the other parameter
Catalog No. NC2741115