Learn More
TARGETMOL CHEMICALS INC POZIOTINIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T263010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Poziotinib (NOV120101)(NOV120101; HM781-36B) is an irreversible HER1/2/4 inhibitor (IC50s 3/5/23 nM). purity: 99%
Catalog No. 50-257-7077