Learn More
Accuflow Systems 1.5" x 3/4" TC Polypropylene Concentric Reducers
Supplier:  Accuflow Systems PPCR150075
Encompass_Preferred
1.5" x 3/4" TC Polypropylene Concentric Reducers are designed for use in GMP Manufacturing and laboratories. These Reducers can be autoclavable as well as gama irradiated. Constructed of USP Class VI Polypropylene. Designed for Sanitary Tri clamp applications.
Catalog No. 50-110-8509