Learn More
Selleck Chemical LLC PP 3 E2980-5MG
SDP
Supplier:  Selleck Chemical LLC E29805MG
Encompass_Preferred
4-AMino-1-phenylpyrazole[3 4-d]pyriMidine(PP 3) is an EGFR tyrosine kinase inhibitor with an IC50 of 2 7 M It is a negative control for the Src kinase inhibitor PP 2
Catalog No. 50-000-40434