Learn More
Bellco Glass, Inc. Polypropylene Erlenmeyer Grad Flask,250mL
SDP
Supplier:  Bellco Glass, Inc. 250000250
Encompass
Flask have wide-mouth design for easy pouring and printed scale of approximate volumes Material: polypropylene Includes: polypropylene screw cap Size: 250ml Volume: 250 x 50mm Thread: 52mm
Catalog No. 50-194-5433