Learn More
MSC Vikan Tank Brush, Stiff Polyester Bristles, Polypropylene Block, FDA Approved
Supplier:  MSC 47177019
Encompass
This oval-shaped tank brush is designed for effective scrubbing inside large industrial tanks, vats, and channel drains. It features stiff polyester bristles and a durable polypropylene block, making it suitable for demanding cleaning tasks in various environments. The brush is made from FDA-compliant materials and is autoclavable and dishwasher safe for easy sanitation.
  • Stiff polyester bristles for effective cleaning
  • Polypropylene block construction
  • Suitable for large industrial tanks, vats, and channel drains
  • Made from FDA-compliant materials
  • Autoclavable for sterilization
  • Dishwasher safe for easy cleaning
  • Staple set construction
  • Supports color-coding programs to reduce cross-contamination
Catalog No. 50-283-5519