Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Vikan Meat Mincer Brush, Stiff, Polyester, 5.00 in, Yellow

Supplier:  MSC 47173679

Encompass

This Meat Mincer Brush features stiff polyester bristles and a 5.00 in diameter, designed for effective cleaning in industrial or commercial meat processing equipment.

Features and Benefits:

  • Stiff bristle flexibility for effective scrubbing
  • Durable polyester bristle material
  • Bright yellow bristle color for easy identification
  • Diameter of 5.00 in
  • Head length of 2.88 in
  • Manufactured in Denmark

Catalog No. 50-283-8170


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.