Learn More
Thomas Scientific PP RESERVOIR PYRAMID BASE25/CS
SDP
Supplier:  Thomas Scientific 1149Q42
Encompass
4 column polypropylene reservoir, 73ml/col, 292ml max, pyramid base geometries, 44mm height
Catalog No. NC2127433