Learn More
AVANTOR MASTERFLEX (VWR LLC) Plastic sanitary couplings; tubing ID, 1/4""; PP; 1 9/16"" x 31/32"" x 3/16""
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3151200
Encompass
Tapered funnel design reduces "dead space" for exceptionally smooth fluid flow. Smooth inner surface prevents particle entrapment; seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth. Sterilize by gamma irradiation. Choose from three polymers; order silicone gasket and push/pull clamp separately below.
Catalog No. NC9520197