Learn More
AdipoGen PP1 (5 mg)
SDP
Supplier:  AdipoGen AGCR13562M005
Encompass_Preferred
Chemical. CAS: 172889-26-8. Formula: C16H19N5. MW: 281.4. Highly potent and selective Scr family tyrosine kinase inhibitor. Shows anti-Ras cancer potential by blocking Ras-induced activation of PAK1. Antitumor compound. RIP2 inhibitor. Blocks TGF-beta-mediated cellular responses.
Catalog No. 50-205-3929