Learn More
Cayman Chemical PP1 (Src Inhibitor), 172889-26-8, 5 mg
SDP
Supplier:  Cayman Chemical 142445
Encompass_Preferred
A potent, reversible, ATP-competitive, and selective inhibitor of the Src family of protein tyrosine kinases: p56lck (IC50 = 5 nM), p59fynT (IC50 = 6 nM), Hck (IC50 = 20 nM)
Catalog No. NC1371100