Learn More
TARGETMOL CHEMICALS INC PP121 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2415
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PP-121 is a multi-targeted inhibitor of PDGFR (IC50 2 nM) Hck (IC50 8 nM) mTOR (IC50 10 nM) VEGFR2(IC50 12 nM) Src (IC50 14 nM) and Abl (IC50 18 nM) also inhibits DNA-PK (IC50 60 nM). purity: 99%
Catalog No. 50-258-0828