Learn More
Sino Biological Peptide Substrates: PP2B Substrate
Supplier:  Sino Biological P51581000
Encompass_Preferred
The PP2B Substrate peptide [DLDVPIPGRFDRRV(p)SVAAE] is derived from bovine PRKAR2A (cAMP-dependent protein kinase type II-alpha regulatory subunit) (amino acids 82-100) and is suitable as PP2B substrate.
Catalog No. 50-256-8185