Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pp60 c-src (521-533) (phosphorylated) | 149299-77-4 | 98.8% | 1543.48 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYP11991MG

Encompass_Preferred

pp60 c-src (521-533) (phosphorylated) is a peptide corresponding to the pp60c-src carboxy terminal regulatory domain. This product is phosphorylated at Tyr527 and is intended for research applications.

  • Peptide corresponding to pp60c-src carboxy terminal regulatory domain
  • Phosphorylated at Tyr527
  • Molecular weight of 1543.48
  • CAS number 149299-77-4
  • Purity of 98.8%
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-24516


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.