Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PPA | 452072-22-9 | 99.0% | 243.35 | C10H13NO2S2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY141664100MG

Encompass_Preferred

PPA is an antibody-drug conjugate (ADC) linker intended for research use. It is used to synthesize ADCs and is supplied with reported high purity and defined storage recommendations.

  • High purity (99.0%)
  • Cas number 452072-22-9
  • Formula C10H13NO2S2
  • Molecular weight 243.35 g/mol
  • Available in multiple solid package sizes, including 100 MG
  • Stored sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month)
  • For research use only; not for human therapeutic use

Catalog No. 50-002-03989


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.