Learn More
ABclonal Technology PPA2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A158191000UL
Encompass_Preferred
The protein encoded by this gene is localized to the mitochondrion, is highly similar to members of the inorganic pyrophosphatase (PPase) family, and contains the signature sequence essential for the catalytic activity of PPase. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Catalog No. 50-001-08743