Learn More
Haematologic Technologies Inc PPACK 25MG
Supplier:  Haematologic Technologies Inc FPRCK0125MG
Encompass_Preferred
Inhibitor, Phe-Pro-Arg Chloromethylketone (PPACK), 25 milligrams, Use: Tri-peptide chloromethylketones are used to irreversibly inhibit various serine proteases, Research use only, Supplied lyophilized, store at 4 degrees Celsius.
Catalog No. 50-883-395