Learn More
Medchemexpress LLC HY-101044 5mg Medchemexpress, PPADS (tetrasodium) CAS:192575-19-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1010445MG
Encompass_Preferred
Medchemexpress, HY-101044 5mg PPADS (tetrasodium) CAS:192575-19-2 PPADS tetrasodiuma is a non-selective P2X receptor antagonist. PPADS tetrasodiuma blocks recombinant P2X1, -2, -3, -5 with IC50s ranging from 1 to 2.6 μM. PPADS tetrasodiuma blocks native P2Y2-like (IC50~0.9 mM) and recombinant P2Y4 (IC50~15 mM) receptors. PPADS tetrasodiuma is an inhibitor of the reverse mode of the Na/Ca²⁺exchanger in guinea pig airway smooth muscle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7801