Learn More
Proteintech Group Inc PPAT Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 154011AP150UL
Encompass_Preferred
The PPAT antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PPAT. This antibody recognizes human, mouse, rat antigen. The PPAT antibody has been validated for the following applications: IP, WB, ELISA analysis.
Catalog No. 50-225-6401