Learn More
Medchemexpress LLC PPM-3 | 3032388-42-1 | 98.2% | 1000.26 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1496165MG
Encompass_Preferred
PPM-3 is a potent and selective PROTAC ERK5 degrader with an IC50 of 62.4 nM. It does not directly influence tumor cell growth but affects tumor development by influencing the differentiation of macrophages.
  • Potent and selective PROTAC ERK5 degrader
  • IC50 of 62.4 nM
  • Affects tumor development by influencing macrophage differentiation
  • Initiates ERK5 degradation within 2-4 hours, with peak degradation at 12 hours
  • Maintains ERK5 degradation for at least 72 hours
  • Shows ERK5 degradation activity in multiple cancer cell lines including H1975, HepG2, MDA-MB-231, PC-3, HCT116, and A375
Catalog No. 50-003-60296