Learn More
Selleck Chemical LLC PPQ-102 S0198-25MG
SDP
Supplier:  Selleck Chemical LLC S019825MG
Encompass_Preferred
PPQ-102 (CFTR Inhibitor IV) is a potent inhibitor of CFTR PPQ-102 can completely inhibit CFTR chloride current with IC50 of 90 nM
Catalog No. 50-000-39605