Learn More
Enzo Life Sciences PQ401 (5mg). CAS: 196868-63-0
SDP
Supplier:  Enzo Life Sciences ALX270459M005
Encompass_Preferred
Potent inhibitor of insulin-like growth factor I receptor (IGF-IR) signalling and breast cancer cell growth in culture and in vivo. Alternative name: IGF-IR Inhibitor, N-(5-chloro-2-methoxyphenyl)-N'-(2-methyl-4-auinolinyl)urea. Purity: ≥98% (HPLC (UV)). Appearance: Off-white solid. Solubility: Soluble in DMSO (10mg/ml) or methanol (7mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8581