Learn More
Medchemexpress LLC PqsR/LasR-IN-1 | 924818-33-7 | 98.1% | 405.87 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY146327
Encompass_Preferred
PqsR/LasR-IN-1 is a potent inhibitor of PqsR and LasR systems. It demonstrates anti-virulence activity against *Pseudomonas aeruginosa*, reducing the production of biofilm, pyocyanin, and rhamnolipids in PA.
  • Potent PqsR and LasR systems inhibitor
  • Exhibits anti-virulence activity against *Pseudomonas aeruginosa*
  • Reduces production of biofilm in PA
  • Reduces production of pyocyanin in PA
  • Reduces production of rhamnolipids in PA
Catalog No. 50-003-63454