Learn More
Medchemexpress LLC PqsR/LasR-IN-3 | 2581109-51-3 | 311.35 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14632950MG
Encompass_Preferred
PqsR/LasR-IN-3 (Compound 7a) is a potent inhibitor of PqsR and LasR systems in *P. aeruginosa*. This compound acts as an antibiotic, targeting bacterial pathways for anti-infection purposes.
  • Potent inhibitor of PqsR and LasR systems in *P. aeruginosa*
  • Inhibits hERG with an IC₅₀ of 109.01 μM
  • Acts as an antibiotic
  • Targets bacterial pathways for anti-infection
Catalog No. 50-003-63833