Learn More
Apexbio Technology LLC PR-619, 25mg. Cas: 2645-32-1 MFCD: MFCD00830384. Deubiquitylating enzymes (DBUs) inhibitor
SDP
Supplier:  Apexbio Technology LLC A821225
Encompass_Preferred
Cell-permeable, non selective, reversible deubiquitinases/deubiquitylases (DUBs) inhibitor 2645-32-1. MFCD00830384
Catalog No. 50-101-1330