Learn More
Medchemexpress LLC PR-619 | 2645-32-1 | 99.26% | 223.28 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13814100MG
Encompass_Preferred
PR-619 is a broad-range and reversible DUB inhibitor with EC50s of 3.93, 4.9, 6.86, 7.2, and 8.61 μM for USP4, USP8, USP7, USP2, and USP5, respectively. It induces ER Stress and ER-Stress related apoptosis.
  • Broad-range and reversible DUB inhibitor
  • Induces ER Stress and ER-Stress related apoptosis
  • Prevents degradation, suggesting KCa3.1 is targeted for degradation by ubiquitylation
  • Affects the microtubule network and leads to the accumulation of small punctuated tau deposits
  • Causes the dephosphorylation of tau
  • Induces heat shock proteins and an increase of ubiquitinated proteins
  • Induces G0/G1 arrest in UC cells
  • Enhances the antitumor effect of Cisplatin in vivo
Catalog No. 50-003-51258