Learn More
Abcam PR 619, DUB inhibitor, 5MG
Supplier:  Abcam AB1446415MG
Encompass_Preferred
MW 223.3 Da, Purity >99%. Cell-permeable, non-selective and reversible DUB inhibitor (IC₅₀ values are 5 to 20 μM). Induces polyubiquitinated protein accumulation in cells without directly affecting proteasome activity. Activates autophagy.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5535