Learn More
Medchemexpress LLC HY-N0082 5mg Medchemexpress, Praeruptorin B CAS:73069-28-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN00825MG
Encompass_Preferred
Medchemexpress, HY-N0082 5mg Praeruptorin B CAS:73069-28-0 Praeruptorin B is an inhibitor of sterol regulatory element-binding proteins (SREBPs) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3381