Learn More
Medchemexpress LLC PR5-LL-CM01 | 1005307-86-7 | 98.4% | 401.51 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY109963
Encompass_Preferred
PR5-LL-CM01 is a potent protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 7.5 μM. It exhibits anti-tumor activities and is intended for research use only.
  • Potent PRMT5 inhibitor (IC50 7.5 μM)
  • Exhibits anti-tumor activities
  • Higher efficacy in inhibiting cancer cells
  • Low toxicity in normal cells
  • Strongly inhibits colony-forming ability in PANC1 and HT29 cells
  • Inhibits NF-κB activation and target gene expression
  • Dramatically decreases TNFα and IL8 expression
Catalog No. 50-003-65757