Learn More
Apexbio Technology LLC Pracinostat (SB939), 5mg. Cas: 929016-96-6 MFCD: MFCD17215206. Pan-HDAC inhibitor
SDP
Supplier:  Apexbio Technology LLC A40955
Encompass_Preferred
Pracinostat, also known as SB939, is a potent and orally available inhibitor of histone deacetylase (HDAC) with a relatively stronger selectivity (more than 1000-fold) for class I, class II and class IV HDACs rather than class III HDACs. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4877