Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Praeruptorin A | 73069-27-9 | MFCD31561439 | 99.6% | 386.40 | C21H22O7 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN606510MG

Encompass_Preferred

Praeruptorin A is a natural coumarin constituent used as a research reagent and analytical standard with reported anti-inflammatory activity via NF-κB inhibition.

  • Pack contains 10 mg of Praeruptorin A.
  • High purity (99.57%).
  • Solid, white to off-white appearance.
  • Molecular formula C21H22O7, molecular weight 386.40.
  • CAS number 73069-27-9.
  • Store at 4°C and protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Intended for in vitro research and analytical applications.

Catalog No. 50-002-89748


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.