Learn More
Medchemexpress LLC Praeruptorin B | 73069-28-0 | MFCD09839133 | 100.0% | 426.46 | C24H26O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN008210MG
Encompass_Preferred
Praeruptorin B is a natural product coumarin used as a research reagent that inhibits sterol regulatory element-binding proteins (SREBPs) and modulates lipid and fatty acid synthesis. Supplied as a white to off-white solid, it is intended for laboratory research applications.
Catalog No. 50-002-95651