Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Praeruptorin E | 78478-28-1 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN60661MG

Encompass_Preferred

Praeruptorin E is a primary bioactive constituent found in Peucedanum praeruptorum, also known as Bai-Hua Qian Hu. It acts as a calcium antagonist, with a pD2′ value of 5.2. This product is intended for research use only and is not sold to patients.

  • High purity: 99.18% (data sheet) or 99.83% (select batch)
  • Solid, white to off-white appearance
  • Ships at room temperature (continental US); store at -20°C, protect from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protect from light
  • Soluble in DMSO at 25 mg/mL (58.35 mM) with ultrasonication
  • Can be prepared with 10% DMSO and 90% corn oil for in vivo studies, yielding a clear solution of ≥ 2.5 mg/mL

Catalog No. 50-004-20091


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.