Learn More
Medchemexpress LLC Praliciguat | 1628730-49-3 | MFCD32062661 | 99.4% | 534.36 g/mol | C21H14F8N6O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1090395MG
Encompass_Preferred
Praliciguat (IW-1973) is a potent, orally active soluble guanylate cyclase (sGC) stimulator supplied as a research-grade small-molecule powder for in vitro and in vivo studies of metabolic and cardiovascular disease. Intended for research use only; follow laboratory safety and storage recommendations.
Catalog No. 50-002-64423