Learn More
Selleck Chemical LLC Pralidoxime chloride S4575-100mg
SDP
Supplier:  Selleck Chemical LLC S4575100MG
Encompass_Preferred
Pralidoxime chloride (2-PAM) is an antidote to organophosphate pesticides and chemicals An acetylcholinesterase (AChE) reactivator
Catalog No. 50-311-1498